ENA018108882 Enamine

4-(chloromethyl)-7-methyl-6-(propan-2-yl)-2H-chromen-2-one

  • MDL number MFCD03978447
  • Cas Number : 568555-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05623
Availability details for
Quantity
  • Availability
  • Add to Cart