ENA018108886 Enamine

pyrrolidine-1-carboxamide

  • MDL number MFCD00509172
  • Cas Number : 4736-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221-224
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05627
Availability details for
Quantity
  • Availability
  • Add to Cart