ENA018108896 Enamine

3-methyl-2-sulfanyl-5-(thiophen-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD06654929
  • Cas Number : 257870-39-6
  • Molecular Weight : 280.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-05637

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05637
Availability details for
Quantity
  • Availability
  • Add to Cart