ENA018108900 Enamine

2-amino-7-methyl-5H-[1,3,4]thiadiazolo[3,2-a]pyrimidin-5-one

  • MDL number MFCD00466623
  • Cas Number : 31737-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-226
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05641
Availability details for
Quantity
  • Availability
  • Add to Cart