ENA018108906 Enamine

[(2-methoxyphenyl)methyl]thiourea

  • MDL number MFCD03978472
  • Cas Number : 66892-30-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05647
Availability details for
Quantity
  • Availability
  • Add to Cart