ENA018108925 Enamine

2-(3-ethylphenoxy)acetic acid

  • MDL number MFCD03980935
  • Cas Number : 1878-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-72
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05667
Availability details for
Quantity
  • Availability
  • Add to Cart