ENA018108939 Enamine

5-Dimethylsulfamoyl-2-pyrrolidin-1-yl-benzoic acid

  • MDL number MFCD03980947
  • Cas Number : 565171-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05681

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05681
Availability details for
Quantity
  • Availability
  • Add to Cart