ENA018108949 Enamine

3-methyl-6-phenyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD06380338
  • Cas Number : 851398-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05693

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05693
Availability details for
Quantity
  • Availability
  • Add to Cart