ENA018108964 Enamine

2-(2,3-dihydro-1,4-benzodioxine-6-carbonyl)benzoic acid

  • MDL number MFCD03982033
  • Cas Number : 568555-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-146
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05708
Availability details for
Quantity
  • Availability
  • Add to Cart