ENA018108967 Enamine

2-(2-chloroacetamido)-N-(4-methoxyphenyl)benzamide

  • MDL number MFCD03987986
  • Cas Number : 22312-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05711
Availability details for
Quantity
  • Availability
  • Add to Cart