ENA018108968 Enamine

2-(2-Chloro-acetylamino)-N-(4-methyl-benzyl)-benzamide

  • MDL number MFCD03982576
  • Cas Number : 736977-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05712

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05712
Availability details for
Quantity
  • Availability
  • Add to Cart