ENA018108970 Enamine

2-Amino-5,5,7,7-tetramethyl-4,5,6,7-tetrahydro-thieno[2,3-c]pyridine-3-carbonitrile

  • MDL number MFCD00499047
  • Cas Number : 133894-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197...199
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05714

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05714
Availability details for
Quantity
  • Availability
  • Add to Cart