ENA018108971 Enamine

(2,5-Dimethyl-benzenesulfonylamino)-acetic acid

  • MDL number MFCD03658020
  • Cas Number : 13240-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05715

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-05715
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart