ENA018109020 Enamine

5-methyl-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD03161246
  • Cas Number : 31130-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-35
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05778
Availability details for
Quantity
  • Availability
  • Add to Cart