ENA018109025 Enamine

6-amino-1-(2-methylpropyl)-5-[(2-methylpropyl)amino]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03982100
  • Cas Number : 571150-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05784
Availability details for
Quantity
  • Availability
  • Add to Cart