ENA018109034 Enamine

3-ethoxy-4-(2-methylpropoxy)benzaldehyde

  • MDL number MFCD03982042
  • Cas Number : 93567-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 21-22
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05797
Availability details for
Quantity
  • Availability
  • Add to Cart