ENA018109039 Enamine

(Tetrahydro-furan-2-ylmethyl)-urea

  • MDL number MFCD03982028
  • Cas Number : 38336-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 75...77
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05802

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05802
Availability details for
Quantity
  • Availability
  • Add to Cart