ENA018109045 Enamine

2-chloro-1-(3,4-difluorophenyl)propan-1-one

  • MDL number MFCD04610455
  • Cas Number : 561009-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 30-31
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05809
Availability details for
Quantity
  • Availability
  • Add to Cart