ENA018109051 Enamine

5-(5-chloro-2-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03983038
  • Cas Number : 554423-96-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05815
Availability details for
Quantity
  • Availability
  • Add to Cart