ENA018109063 Enamine

N-(2-chloroacetyl)-4-(difluoromethoxy)benzamide

  • MDL number MFCD03982152
  • Cas Number : 568555-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05829
Availability details for
Quantity
  • Availability
  • Add to Cart