ENA018109077 Enamine

ethyl 2-amino-5-carbamoyl-4-methylthiophene-3-carboxylate

  • MDL number MFCD00458467
  • Cas Number : 43028-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05846
Availability details for
Quantity
  • Availability
  • Add to Cart