ENA018109086 Enamine

2-(2-chloroacetamido)-N-[2-(4-methoxyphenyl)ethyl]benzamide

  • MDL number MFCD04614032
  • Cas Number : 117507-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-136
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05859
Availability details for
Quantity
  • Availability
  • Add to Cart