ENA018109090 Enamine

7-methyl-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD01648761
  • Cas Number : 41177-66-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05864
Availability details for
Quantity
  • Availability
  • Add to Cart