ENA018109097 Enamine

4-Morpholin-4-yl-[1,2,5]thiadiazol-3-ol

  • MDL number MFCD01820806
  • Cas Number : 30165-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192...194
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05872

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05872
Availability details for
Quantity
  • Availability
  • Add to Cart