ENA018109098 Enamine

1-(4-fluorobenzenesulfonyl)-4-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD03982078
  • Cas Number : 1008962-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05874
Availability details for
Quantity
  • Availability
  • Add to Cart