ENA018109108 Enamine

N-(6-amino-1-benzyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)-2-chloro-N-(2-methylpropyl)acetamide

  • MDL number MFCD04610476
  • Cas Number : 561009-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05887
Availability details for
Quantity
  • Availability
  • Add to Cart