ENA018109109 Enamine

N-(6-amino-1-benzyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)-2-chloro-N-propylacetamide

  • MDL number MFCD04610462
  • Cas Number : 571917-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05888

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05888
Availability details for
Quantity
  • Availability
  • Add to Cart