ENA018109113 Enamine

5-(4-Dimethylamino-benzylidene)-2-thioxo-imidazolidin-4-one

  • MDL number MFCD00450950
  • Cas Number : 41250-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05893

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05893
Availability details for
Quantity
  • Availability
  • Add to Cart