ENA018109116 Enamine

2-(2-chloroacetamido)-N-(4-ethoxyphenyl)benzamide

  • MDL number MFCD03986472
  • Cas Number : 565174-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05896
Availability details for
Quantity
  • Availability
  • Add to Cart