ENA018109131 Enamine

3-(benzylsulfanyl)-1,2,4-thiadiazol-5-amine

  • MDL number MFCD00649535
  • Cas Number : 83757-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-110
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05914
Availability details for
Quantity
  • Availability
  • Add to Cart