ENA018109134 Enamine

5-cyclohexyl-1-phenyl-1H-1,2,4-triazole-3-thiol

  • MDL number MFCD04610447
  • Cas Number : 571917-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05918

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05918
Availability details for
Quantity
  • Availability
  • Add to Cart