ENA018109140 Enamine

2-Difluoromethoxy-benzaldehyde oxime

  • MDL number MFCD01124382
  • Cas Number : 332110-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05924

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05924
Availability details for
Quantity
  • Availability
  • Add to Cart