ENA018109141 Enamine

N-(2,3-dimethylphenyl)-2-(N-hydroxyimino)acetamide

  • Cas Number : 6579-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-111
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05925
Availability details for
Quantity
  • Availability
  • Add to Cart