ENA018109146 Enamine

3-(1H-indol-3-yl)-3-oxopropanenitrile

  • MDL number MFCD04610443
  • Cas Number : 20356-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05930
Availability details for
Quantity
  • Availability
  • Add to Cart