ENA018109151 Enamine

N-(2,3-dihydro-1,4-benzodioxin-6-yl)-2-(N-hydroxyimino)acetamide

  • Cas Number : 154869-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05935
Availability details for
Quantity
  • Availability
  • Add to Cart