ENA018109155 Enamine

5-(piperidine-1-sulfonyl)-2-(2,2,2-trifluoroethoxy)aniline

  • MDL number MFCD04610255
  • Cas Number : 556016-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05941

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05941
Availability details for
Quantity
  • Availability
  • Add to Cart