ENA018109163 Enamine

2-Mercapto-benzooxazole-5-sulfonic acid (2-chloro-phenyl)-amide

  • MDL number MFCD03150432
  • Cas Number : 380426-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05950

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05950
Availability details for
Quantity
  • Availability
  • Add to Cart