ENA018109164 Enamine

3-(4-amino-5-sulfanyl-4H-1,2,4-triazol-3-yl)-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD03985192
  • Cas Number : 571154-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05951
Availability details for
Quantity
  • Availability
  • Add to Cart