ENA018109168 Enamine

5-ethoxy-3-methyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD03985193
  • Cas Number : 568558-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-156
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05955
Availability details for
Quantity
  • Availability
  • Add to Cart