ENA018109174 Enamine

methyl 5-[(2-ethoxy-4-formylphenoxy)methyl]furan-2-carboxylate

  • MDL number MFCD04610463
  • Cas Number : 733796-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05961
Availability details for
Quantity
  • Availability
  • Add to Cart