ENA018109180 Enamine

4-(2,4,5-trimethylphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02663890
  • Cas Number : 383131-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05970
Availability details for
Quantity
  • Availability
  • Add to Cart