ENA018109201 Enamine

6-(Piperidine-1-sulfonyl)-benzotriazol-1-ol

  • MDL number MFCD03985190
  • Cas Number : 733762-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05998

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05998
Availability details for
Quantity
  • Availability
  • Add to Cart