ENA018109205 Enamine

5-nitro-1H-1,2,3-benzotriazol-1-ol

  • MDL number MFCD00760413
  • Cas Number : 64352-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06003
Availability details for
Quantity
  • Availability
  • Add to Cart