ENA018109208 Enamine

3-[(3,5-dimethyl-1,2-oxazol-4-yl)methoxy]benzoic acid

  • MDL number MFCD02324525
  • Cas Number : 315692-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06007
Availability details for
Quantity
  • Availability
  • Add to Cart