ENA018109212 Enamine

N-(4-methoxy-3-sulfamoylphenyl)acetamide

  • MDL number MFCD03983099
  • Cas Number : 568556-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06011
Availability details for
Quantity
  • Availability
  • Add to Cart