ENA018109220 Enamine

3-[benzyl(phenyl)sulfamoyl]-4-chlorobenzoic acid

  • MDL number MFCD03985224
  • Cas Number : 733762-51-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06020
Availability details for
Quantity
  • Availability
  • Add to Cart