ENA018109228 Enamine

3-ethoxy-4-(2-methylpropoxy)benzoic acid

  • MDL number MFCD03985219
  • Cas Number : 565173-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06029
Availability details for
Quantity
  • Availability
  • Add to Cart