ENA018109231 Enamine

2-chloro-N-[2-(2,4-dichlorophenyl)pentyl]acetamide

  • MDL number MFCD04610441
  • Cas Number : 733796-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06033
Availability details for
Quantity
  • Availability
  • Add to Cart