ENA018109244 Enamine

3-(1-phenylethyl)-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD03985197
  • Cas Number : 568558-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06051
Availability details for
Quantity
  • Availability
  • Add to Cart