ENA018109251 Enamine

4-butoxy-N-hydroxy-3-methoxybenzamide

  • MDL number MFCD03985236
  • Cas Number : 565173-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-06059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06059
Availability details for
Quantity
  • Availability
  • Add to Cart